Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RS Urine Sediment Workstation, Apacor Limited

Supplier: Apacor

The RS Urine Sediment Workstation is designed for the preparation of samples for urine sediment analysis.

The Workstation operates in conjunction with any standard, upright microscope. Sample is mixed and loaded automatically onto the “Optical Slide Assembly” (OSA). Viscous samples are easily diluted to provide a useful sample.

There is no sample migration to the outside of the coverslip, as the sample “floats” within the OSA. The sampling process is fully enclosed with no mess. There are no slides or coverslips required. The OSA performs this job, and is reusable.

Ordering Information: The RS Urine Sediment Workstation comes complete with one optical slide assembly without grid, aspirator kit (includes aspirator probe, probe handle, and tubing to connect the aspirator probe to the OSA), tubing kit (includes all user serviceable tubing, pre-cut and pre-fitted), purge bottle, bleach bottle, 48-position tube rack, power supply unit, and one-year manufacturer's warranty.

CE certified.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antibody Type Primary Control
Antigen Name Breast Cancer type 1 susceptibility protein Immunoglobulin G1 HA Epitope Tag Actin Beta
Antigen Synonyms BRCAI, BRCC1, BROVCA1, IRIS, PNCA4, PPP1R53, PSCP, RNF53, BRCA1/BRCA2-containing complex, subunit 1, RING finger protein 53, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein 1, breast cancer type 1 susceptibility protein, protein phosphatase 1, regulatory subunit 53 human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag BRWS1, PS1TP5BP1, PS1TP5-binding protein 1, actin, cytoplasmic 1, beta cytoskeletal actin
ChIP Yes
Clonality Monoclonal
Clone MS110 2-2.2.14 15G5A11/E2
Conjugation Unconjugated Biotin
Cross Adsorption No
ELISA Yes
Flow Cytometry Yes
Form Liquid
Gel Shift Yes
Gene ID ACTB/60; Actb/11461/81822
Host Mouse
ImmunoChemistry Yes
ImmunoFluorescence Yes
ImmunoPrecipitation Yes
Isotype IgG1
Neutralisation Yes
Reactivity Human Tags Rat, Mouse, Human, Primates
Storage Buffer PBS with 1mg/ml BSA, 30% glycerol and 0.05% Sodium azide PBS with 1% BSA and 0.09% Sodium azide PBS, pH 7.4, with 0.05% Sodium azide
Storage Temperature -20°C 4°C or -20°C if preferred -20° C, Avoid Freeze/Thaw Cycles
Western Blot Yes

ACCESSORIES

Expand All / Collapse All

Accessories
Description Details Supplier No. VWR Catalog Number Unit Price Quantity
Bleach Bottle - 90033033000 10144-696 Each Retrieving Restricted
Optical Slide Assembly without Grid - 900110190002 10144-714 Each Retrieving Restricted
Urizyme® Enzymatic Detergent An anionic powdered detergent for use as a cleansing agent for the RSWorkstation's flow cells (housed within the Workstation's
Optical Slide Assembly); An odourless powder that aids in reducing protein build up within the flow cells and is soluble in water making it very simple and convenient to use; Protease enzyme removes tissue, blood and body fluids; Biodegradable and readily disposable; CE certified; Plastic Vial.
1699 10200-132 Box of 30 Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty