Would you like to log in again?
KWIK-STIK Escherichia coli ATCC® 23848™, Microbiologics
Supplier: MICROBIOLOGICS, INC.
0203P 0203K
The KWIK-STIK™ is well-known for its clever design and accurate, repeatable results
Each KWIK-STIK™ device features a single QC microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid, and an inoculating swab. For added convenience, a peel-off identification label is included for easy documentation.
Certifications: The KWIK-STIK™ falls under the category of In-Vitro Diagnostics Medical Devices Directives for CE Mark Conformity. This product is also listed with the FDA.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the vwr.com product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.
ORDER
Expand All / Collapse All
Description Culture Packaging Supplier No. VWR Catalog Number Unit Price Quantity
KWIK-STIK Escherichia coli ATCC® 23848™* Pack of 2 0203P 89502-870 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
KWIK-STIK Escherichia coli ATCC® 23848™* Pack of 6 0203K 89502-866 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Certification Online Certificate of Analysis - Documentation of phenotypic, macroscopic and microscopic test results
Traceability Assured Quality - Traceable to reference culture
Storage Conditions Easy storage; no dry ice required; reduces shipping costs
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty