Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Thermo Scientific (14) Aes Chemunex Inc (1)
  • Search Within Results
You Searched for: "Microplate Dispensers"
https://us.vwr.com/stibo/smallweb/std.lang.all/53/28/18945328.jpg
High-speed microplate dispensers precisely deliver desired volumes rapidly and uniformly. Extremely adaptable, the equipment can quickly switch to function with different sized multi-well plates. Easily adjust operations using the simple interface settings and graphic display. With a compact design, microplate dispensers use limited benchtop space. Detachable, autoclavable parts eliminate testing contamination. Black-flushing models push expensive reagents back in to the bottles to minimize unnecessary loss. Multiple interior channel styles dispense different liquids simultaneously.
SearchPresentationType-VERTICAL
SearchResultCount:"5"
Sort by:
Supplier:  Thermo Scientific
Description:   Multidrop Combi Reagent Dispenser, Thermo Scientific
Supplier:  Thermo Scientific
Description:   Multidrop* 384 Reagent Dispenser
Supplier:  Thermo Scientific
Description:   Multidrop* DW Reagent Dispenser
Supplier:  Aes Chemunex Inc
Description:   Rrevolutionnary Culture Media Dispensing System
Supplier:  Thermo Scientific
Description:   MULTIDROP COMBI NL (100-240 V, 50/60 Hz)
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 5  of 5