Sulfuric acid 0.0200 N (N/50) in aqueous solution

Supplier: Ricca Chemical

8200-16
RC8200-16EA 32.2 USD
RC8200-16
Sulfuric acid 0.0200 N (N/50) in aqueous solution
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Assay 0.02 ± 0.00002 N at 20 °C
NIST SRM 723

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR