Hydrogen peroxide 30% (w/w) ACS, Reagent Grade
Supplier: Ricca Chemical
3821.7-32
3821.7-1
RC3821.7-1EA
745.22
USD
RC3821.7-1
RC3821.7-32
Hydrogen peroxide 30% (w/w) ACS, Reagent Grade
Hydrogen peroxide
Formula:
H₂O₂ MW: 34.01 g/mol |
MDL Number:
MFCD00011333 CAS Number: 7722-84-1 UN: 2014 ADR: 5.1,II |
Specification Test Results
Residue After Evaporation | 0.00002 % max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...