Hydrochloric acid 3.00 N
Supplier: Ricca Chemical
Synonyms:
Muriatic acid
3720-16
RC3720-16EA
39.1
USD
RC3720-16
Hydrochloric acid 3.00 N
Hydrochloric acid
Formula:
HCl MW: 36.46 g/mol |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 UN: 1789 ADR: 8,II |
Specification Test Results
Assay | 3 ± 0.001 N at 20 °C |
NIST SRM | 84 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...