Benedict's reagent quantitative

Supplier: Ricca Chemical

935-16
RC935-16EA 77.12 USD
RC935-16
Benedict's reagent quantitative
Benedict's reagent
MDL Number: MFCD00146068
CAS Number: 63126-89-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 2 ± 0.01 mg Glucose/ mL

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR