Aerosol OT® 10% (w/v)

Supplier: Ricca Chemical

Synonyms: Dioctyl sulfosuccinate sodium saltDocusate sodiumDioctyl sulphosuccinate sodium salt

281-32
RC281-32EA 60.74 USD
RC281-32
Aerosol OT® 10% (w/v)
Aerosol OT®
Formula: C₂₀H₃₇NaO₇S
MW: 444.57 g/mol
MDL Number: MFCD00012455
CAS Number: 577-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR