Salinity/Temperature Meter, Extech
Supplier: Extech
EC170
10111-918EA 147.61 USD
10111-918
Salinity/Temperature Meter, Extech
Electrochemical Meters Salinity Meters
Waterproof Salinity/Temperature meter with high-accuracy multi-ranging sensor
Measure salinity in aquaculture, environmental studies, ground water, Koi ponds, irrigation, drinking water and other applications. Meter features autoranging with two ranges of measurement and a built-in NaCl conductivity to TDS conversion factor.
Certifications: CE