Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

400A AC/DC Clamp Meters + NCV, Extech

Supplier: Extech
MA430T-NIST MA435T MA435T-NIST MA430T MA430
Compact size allows measurements in tight locations

The True RMS AC clamp meter features TRMS measurements, and the True RMS AC/DC clamp meter features TRMS measurements as well as frequency (10Hz to 10kHz), capacitance (100µF or 0.01nF) and temperature (-4 to 1400°F or -20 to 760°C [0.1°]) measurements.

All three units feature a built-in non-contact voltage detector with LED alert and backlit LCD display. The 30 mm (1.2") jaw size fits conductors up to 350 MCM.

Certifications: CE



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
400A AC Clamp Meter + NCV MA430 10111-888 Each Retrieving
400A True RMS AC Clamp Meter + NCV MA430T 10111-890 Each Retrieving
True RMS AC/DC Clamp Meter + NCV MA435T 10111-892 Each Retrieving
400A True RMS AC Clamp Meter + NCV with certificate of calibration traceable to NIST MA430T-NIST 10171-844 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Voltage AC 600V (0.1mV)
Voltage DC 600V (0.1mV)
Current 400A (0.001A) AC, 400A (0.001A) DC 400A (0.001A) AC
Resistance 40MΩ (0.1Ω) 20MΩ (0.1Ω)
Display 4000 counts 2000 counts

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty