Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Invertebrates 27 Piece Embedment Set

Supplier: ED SPELDY EAST CO. LTD.
TE-270
Specimens are preserved and encased in clear resin material.

  • Comparative anatomy
  • Invertebrate classification
  • Diversity of life

There are 27 specimen blocks in this package. They can be observed from every side to see details.

Ordering information: Specimens include chafer beetle (Anomala cuprea hope), lady bug (Synonycha grandis (Thunberg)), mole cricket (Gryllotalpa orientalis Burmeister), praying mantis (Hierodula patellifera (Serville)), wasp (Polistes olivaceus (De Geer)), honey bee (Apis cerana), ant (Pheidologeton latinodus Zhou et Zeng), dung beetle (Catharsius molossus), rhinoceros beetle (Xylotrupes gideon), common tiger butterfly (Danus gentia), long-horned beetle (Anoplophora chinensis), cicada (Cryptoympana atrata), shield bug (Eusthenes cupreus), spider (Argiope bruennichi), dragonfly (Brachythemis contaninata), cricket (Teleogrylius emma), cockroach (Periplaneta australasiae (Fabricius)), scorpion (Mesobuthus martensii), centipede (Scolopendra subspini), walking stick (Gongy10pus adyposus brunner ), onion fly (Delia antiqua Meigen), Chinese shrimp (Penaeus chinensis), silkworm life cycle (Bomombyx mandarina), crab (Eriocheir sinensis), star fish (Asterias amurensis), grasshopper life cycle (Oxya chinensis), and stag beetle (Dorcus titanus platymelus) .Teacher information guide, fact sheets, student activity sheets, and answer keys are included.



 

ORDER

Expand All / Collapse All
Description Height Length Width No. of Parts Supplier No. VWR Catalog Number Unit Price Quantity
Invertebrates 27 Piece Embedment Set 3" 16.5" 13" 27 TE-270 470002-544 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty