Dimethyl carbonate for pharma synthesis, non-GMP, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ400

8.45152.0100 8.45152.0100
EM8.45152.0100CS 737.45 USD
CA8.45152.0100 EM8.45152.0100
Dimethyl carbonate for pharma synthesis, non-GMP, Sigma-Aldrich®
Dimethyl carbonate
Formula: C₃H₆O₃
MW: 90.08 g/mol
MDL Number: MFCD00008420
CAS Number: 616-38-6
UN: 1161
ADR: 3,II

Order Now

Specification Test Results

Solubility 139 g/l (20°C)
Melting Point 0.5 - 4.7 °C
Boiling Point 90 °C (1016 hPa)
Vapor Pressure 53 hPa (20 °C)
Flash Point 16.7 °C
Vapor Pressure 53 hPa (20 °C)
Explosion Limit 4.22 - 12.87 %(V)
Density 1.07 g/cm3 (20 °C)
Ignition Point 458°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR