Dipropylamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
N-Propyl-1-propanamine
M-Clarity™ quality level = MQ200
8.03548.9020
8.03548.9020
EM8.03548.9020EA
1156
USD
CA8.03548.9020
EM8.03548.9020
Dipropylamine for synthesis, Sigma-Aldrich®
Dipropylamine
Formula:
C₆H₁₅N MW: 101.19 g/mol |
MDL Number:
MFCD00009362 CAS Number: 142-84-7 UN: 2383 ADR: 3,II |
Specification Test Results
Solubility | 35 g/l (25°C) (experimental) |
Melting Point | -63 °C |
Boiling Point | 105 °C (1013 hPa) |
Vapor Pressure | 38 hPa (20 °C) |
Flash Point | 7 °C |
Refractive Index | 1.4049 (20 °C, 589 nm) |
Explosion Limit | 1.8 - 9.3 %(V) |
Density | 0.74 g/cm3 (20 °C) |
pH | (1 g/l, H2O, 25 °C) |
Ignition Point | 260°C |
Hill Formula | C6H15N |
Chemical Formula | (CH3CH2CH2)2NH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...