Caproic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Hexanoic acid

M-Clarity™ quality level = MQ200

8.00198.9026 8.00198.9026
EM8.00198.9026EA 1026 USD
CA8.00198.9026 EM8.00198.9026
Caproic acid for synthesis, Sigma-Aldrich®
Caproic acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00004421
CAS Number: 142-62-1
UN: 2829
ADR: 8,III

Order Now

Specification Test Results

Solubility 9.7 g/l (20°C)
Melting Point -3 °C
Boiling Point 206 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 110 °C
Refractive Index 1.4162 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)
pH 4 (1 g/l, H2O, 20 °C)
Ignition Point 380°C
Synonyms Caproic acid
Hill Formula C6H12O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR