EDTA disodium salt dihydrate, Titriplex® III, GR for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Ethylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateEthylenediaminetetraacetic acid, disodium salt, dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIKestranal® 2S dihydrateEdetic acid disodium salt dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrate

1.08418.9025 1.08418.9025
EM1.08418.9025EA 8763 USD
CA1.08418.9025 EM1.08418.9025
EDTA disodium salt dihydrate, Titriplex® III, GR for analysis, Supelco®
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
MDL Number: MFCD00150037
CAS Number: 6381-92-6
https://us.vwr.com/stibo/web/std.lang.all/99/56/2299956.jpg

Order Now

Specification Test Results

Solubility 100 g/l (20°C)
Melting Point 110 °C Elimination of water of crystallisation
Molar Mass 700 kg/m3
Bulk Density 700 kg/m3
pH 4 - 5 (50 g/l, H2O, 20 °C)
Grade ACS,ISO,Reag. Ph Eur
Synonyms Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid disodium salt
Hill Formula C10H14N2Na2O8* 2 H2O
Chemical Formula C10H14N2Na2O8* 2 H2O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR