Legato 110 Single Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
788110
10117-490EA 3755.68 USD
10117-490
Legato 110 Single Syringe Infuse/Withdraw Programmable Syringe Pump, KD Scientific
Infuse/Withdraw/Continuos Single syringe pump
  • Touch pad lock feature, LED light on front panel, USB port, I/O and TTL interface
  • Full metal chassis
  • Built in syringe table, RS485 interface
  • Advanced microstepping techniques
  • Continuous mode of operation
  • Spill dam
  • Programmable
Accomodates syringe sizes 0.5ul to 60 ml. User definable flow rates. High resolutuion touch screen display. Programmable with two user defined programs. Selectable syringe table. Up to 30 lbs of linear force. USB control.
Certifications: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Caution: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.