Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Savant SC210P1 SpeedVac Kit, Thermo Scientific

Supplier: Thermo Scientific
SC210P1-115 SC210P1-230
The Savant SC210 P1 SpeedVac vacuum concentrator kit features an easy-to-use, large capacity concentrator.

  • Long lasting durability against harsh solvents
  • Three selectable heating levels
  • Automatic bleeder valve

The Savant SC210 P1 SpeedVac vacuum concentrator kit features an easy-to-use, large capacity concentrator. Solvents and solvent combinations include acetonitrile, methanol, ethanol and water. This SpeedVac kit is typically used in HPLC fractions with water/0.1% trifluoracetic acid and reversed-phase SPE eluants. Use the complete kit package for HPLC fraction and reversed-phase SPE eluant applications.

The Savant SC210 P1 SpeedVac features a PTFE coated chamber that provides long lasting durability against harsh solvents, three selectable heating levels to optimize evaporation and an automatic bleeder valve that automatically provides vacuum at the beginning of a run and bleeds chamber to atmospheric pressure at the end of a run.

Ordering information: The Savant SC210 P1 SpeedVac kit includes a high-capacity SC210A SpeedVac concentrator, -105°C, ultra-low temperature refrigerated vapor trap, VLP120 vacuum pump, 200 x 1.5 mL rotor (RH200-12), digital vacuum gauge, universal tubing package, chemical trap for use with disposable cartridge, disposable cartridge with color indicator for trapping acids and water vapor, glass condensation flask and CryoCool heat transfer fluid (1 L).



 

ORDER

Expand All / Collapse All
Description Electrical Supplier No. VWR Catalog Number Unit Price Quantity
Savant SC10P1 SpeedVac Kit 115V, 60 Hz SC210P1-115 10081-080 Each Retrieving
Savant SC10P1 SpeedVac Kit 230V, 50 Hz SC210P1-230 10081-100 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty