Savant SPD121P SpeedVac Kit, Thermo Scientific
SPD121PP1-115 SPD121PP1-230
10081-092EA 23339 USD
10081-092 10081-072
Savant SPD121P SpeedVac Kit, Thermo Scientific
The Thermo Scientific Savant SPD121PP1 SpeedVac Kit is reliable, versatile and can be used with a wide variety of solvents.
  • Typically used in removing ethyl acetate from samples
  • Used in drug metabolities
  • Features a deep aluminum PTFE coated chamber
The Thermo Scientific Savant SPD121PP1 SpeedVac Kit is reliable, versatile and can be used with a wide variety of solvents. Solvents and solvent combinations include acetone, chloroform, ethyl acetate, hexane and methylene chloride. This SpeedVac kit is typically used in removing ethyl acetate from samples for CAT assay. It is also used in drug metabolities in solid phase extracts and with analyte elution solvents for evaporation. The Savant SPD121P SpeedVac Kit features a deep aluminum PTFE coated chamber, safety cover lock and a radiant glass safety cover that improves drying time by applying radiant IR heat to samples. Additional features include PTFE plumbing and tubing, two valve manifold with built-in automatic bleeder valve, time/temperature display incorporating membrane touch pad switches, time/vacuum run, variable drying times from 35°C to 80°C in 5 degree increments, manual/auto timed run, endpoint mode that terminates run when pre-set vacuum level is reached, built-in vacuum gauge and vacuum display and standard glass safety lids.
Ordering information: The Savant SPD121P SpeedVac Kit includes the SpeedVac concentrator, -105°C ultra-low temperature refrigerated vapor trap, OFP400 oil free, ETFE/PFA four-headed pump, 64 x 1.5 mL rotor (RH64-11), digital vacuum gauge, chemical trap kit for use with disposable cartridge, disposable cartridge with color indicator for trapping acids and water vapor, tubing and fitting kit including 4' of clear, thick-walled vacuum tubing, two clamps and a cutter, glass condensation flask and CryoCool heat transfer fluid (1 L).