Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Savant SPD1010 and SPD2010 Stand-Alone Vacuum Concentrators, Thermo Scientific

Supplier: Thermo Scientific
SPD1010A-115 SPD2010A-230 SPD1010A-230 SPD2010A-220
SPD1010 and SPD2010 SpeedVac concentrators bring a new level of simplicity to general-purpose vacuum concentration.

  • Fully integrated systems
  • Rapid, efficient concentration of aqueous and mild organic solvent-based samples
  • Ideal solution for busy life science, hospital and government research labs

SPD1010 and SPD2010 SpeedVac concentrators bring a new level of simplicity to general-purpose vacuum concentration. These fully integrated systems support the rapid, efficient concentration of aqueous and mild organic solvent-based samples in biomolecular research applications. Combining versatility and ease of use in a compact footprint, these low-maintenance systems are the ideal solution for busy life science, hospital and government research labs.



 

ORDER

Expand All / Collapse All
Description Electrical Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Savant SPD1010 SpeedVac - Not a Kit 115V, 60 Hz 100 mL SPD1010A-115 10081-106 Each Retrieving
Savant SPD1010 SpeedVac - Not a Kit 230V, 50 Hz 100 mL SPD1010A-230 10081-108 Each Retrieving
Savant SPD2010 SpeedVac - Not a Kit 220V, 60 Hz 500 mL SPD2010A-220 10081-110 Each Retrieving
Savant SPD2010 SpeedVac - Not a Kit 230V, 50 Hz 500 mL SPD2010A-230 10081-112 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty