Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Savant SPD1010 and SPD2010 SpeedVac Kits, Thermo Scientific

Supplier: Thermo Scientific
SPD2010P1-220 SPD1010P1-230 SPD2010P1-230 SPD1010P1-115
These integrated Savant SpeedVac concentrators feature a microprocessor control panel with digital display.

  • Fully incorporate four individual components
  • Typically used in HPLC fractions applications
  • Easily programmable

Solvents and solvent combinations for both the SPD1010 and the SPD2010 include acetonitrile, ethanol, methanol, methylene chloride, water, acetonitrile and water and methanol and water. These integrated Savant SpeedVac concentrators feature a microprocessor control panel with digital display. Both models fully incorporate four individual components. Included are the concentrator bowl, refrigerated trap, vacuum gauge and a diaphragm pump. All-in-one design makes setup and installation easy. These kits are typically used in HPLC fractions applications with water/0.1% trifluoroacetic acid as well as with reveresed-phase SPE eluants. These kits are easily programmable and feature an integrated design for plug-in simplicity and control. Digital display shows time, temperature and vacuum. Vacuum control allows for vacuum ramping and ultimate level setting. This optimizes drying times and eliminates bumping of different solvent mixtures. Dual timers allow for independent control of heater and run time. Oil-free vacuum source eliminates the need to add or change oil. Additional features include a standard PTFE coated chamber, a radiant cover for chemical resistance and added heat source, as well as standard glass safety lids.

Ordering information: The Savant SPD1010 SpeedVac Kit includes the integrated SpeedVac system with oil-free vacuum pump and a -50°C cold trap, RH40-11 40 x 1.5 mL Rotor and CryoCool heat transfer fluid (1 L).
The Savant SPD2010 SpeedVac Kit includes the integrated SpeedVac system with oil-free vacuum pump and a -50°C cold trap, RH200-12 200 x 1.5 mL Rotor and CryoCool heat transfer fluid (1 L).



 

ORDER

Expand All / Collapse All
Description Electrical Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Savant SPD1010 SpeedVac Kit 115V, 60 Hz 100 mL SPD1010P1-115 10098-562 Each Retrieving
Savant SPD1010 SpeedVac Kit 230 V/50 Hz 100 mL SPD1010P1-230 10081-086 Each Retrieving
Savant SPD2010 SpeedVac Kit 220V, 60 Hz 500 mL SPD2010P1-220 10081-078 Each Retrieving
Savant SPD2010 SpeedVac Kit 230 V/50 Hz 500 mL SPD2010P1-230 10081-098 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty