Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SC250EXP Large-Capacity Benchtop SpeedVac Kits, Thermo Scientific

Supplier: Thermo Scientific
SC250P1-230 SC250P1-115 SC250P2-115 SC250P2-230
SC250EXP large-capacity benchtop SpeedVac Kits have everything you need to effectively concentrate high boiling point solvents and strong acids and bases.

  • Drying lipid extracts
  • Complete management of evaporating dissolution solvents
  • Maximum chemical resistance and durability
  • Digital display

The SC250EXP P1 is used in SPE applications, for drying lipid extracts, flash chromatography, solid phase synthesis cleavage solutions such as hydrolyzates and evaporating synthesis solids. Solvents and solvent combinations include acetic acid, acetone, ammonium hydroxide, formic acid, hydrochloric acid, TFA, methylene chloride and water, drug metabolites in SPE/urine, DCM and heptane, methylene chloride and MEOH, THF/pyridine (95:5) and DMSO and methanol. Dual vacuum control provides flexibility to use many solvent types. Four IR heat lamps assist in drying high boiling point solvents. Dual vacuum control of RAMP and LEVEL eliminates bumping or freezing of samples as well as provides optimization for various sample types.

The SC250EXP P2 is used in complete management of evaporating dissolution solvents. Solvents and solvent combinations include chloroform, DMSO, DMF, ethyl acetate, hexane, toluene and methanol and chloroform.

Kalrez® and PTFE coated materials along the entire vapor path allow for maximum chemical resistance and durability. Dual valve mechanism also improves durability and exhaust capabilities. Additional features include an enlarged vacuum port, extra-wide tubing and a new extra-large mouth glass condensation flask. Heated glass lid reduces drying time and prevents condensation. Digital display shows time, temperature and vacuum. Standard glass safety lids are included.

Ordering information: The SC250EXP P1 SpeedVac Kit includes the SC250 concentrator, an ultra-low temperature refrigerated vapor trap (-105°C), VLP120 vacuum pump, FPR4A rotor and 4 UPC1 single shelf microplate carriers, glass condensation flask, tubing kit with 4' clear, thick-walled vacuum tubing, two clamps and a cutter, vacuum pump lubricating oil and 1 L of CryoCool heat trasfer fluid.
The SC250EXP P2 SpeedVac Kit includes the SC250 concentrator, a refrigerated vapor trap (-5°C), VLP120 vacuum pump, FPR4A rotor and 4 UPC1 single shelf microplate carriers, glass condensation flask, tubing kit with 4' clear, thick-walled vacuum tubing, two clamps and a cutter, vacuum pump lubricating oil and 1 L of CryoCool heat trasfer fluid.



 

ORDER

Expand All / Collapse All
Description Electrical Supplier No. VWR Catalog Number Unit Price Quantity
SC250EXP P1 Concentrator 115V, 60 Hz SC250P1-115 10081-066 Each Retrieving
SC250EXP P2 Concentrator 115V, 60 Hz SC250P2-115 10098-560 Each Retrieving
SC250EXP P1 Concentrator 230V, 50Hz SC250P1-230 10081-082 Each Retrieving
SC250EXP P2 Concentrator 230V, 50Hz SC250P2-230 10081-084 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty