Rat Recombinant IFN Gamma (from E. coli)
Supplier: ProSci
40-472
10072-494EA 302.43 USD
10072-494
Rat Recombinant IFN Gamma (from E. coli)
Proteins and Peptides
IFN-gamma is an acid-labile interferon produced by CD4 and CD8 T lymphocytes as well as activated NK cells. IFN-gamma receptors are present in most immune cells, which respond to IFN-gamma signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. IFN-gamma signaling in antigen-presenting cells and antigen-recognizing B and T lymphocytes regulate the antigen-specific phases of the immune response. Additionally, IFN-gamma stimulates a number of lymphoid cell functions including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils. Human IFN-gamma is species-specific and is biologically active only in human and primate cells. Recombinant rat IFN-gamma is a 15.6 kDa protein containing 135 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.