Sodium formate, white powder ACS

Supplier: VWR

Synonyms: Formic acid sodium salt

VWRVJ961-100G
89230-074EA 58.15 USD
89230-074
Sodium formate, white powder ACS
Sodium formate
Formula: HCO₂Na
MW: 68.01 g/mol
Melting Pt: 257.31 °C
Density: 1.919 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 12,08765

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.001%
Heavy Metals = 0.0005%
Insolubles = 0.005%
Iron = 0.0005%
Purity = 99%
Sulfate = 0.001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR