PIPES dipotassium salt, Reagent Grade

Synonyms: Piperazine-N,N'-bis-2-ethanesulphonic acid dipotassium saltDipotassium piperazine-1,4-diethanesulphonatePIPES-K2Dipotassium piperazine-1,4-diethanesulfonate2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid dipotassium saltPiperazine-N,N'-bis-2-ethanesulfonic acid dipotassium saltPiperazine-1,4-bis(2-ethanesulphonic acid) dipotassium salt2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid dipotassium saltPiperazine-1,4-bis(2-ethanesulfonic acid) dipotassium salt

VWRVE912-500G
10118-228EA 34.1 USD
10118-228
PIPES dipotassium salt, Reagent Grade
PIPES dipotassium salt
Formula: C₈H₁₆N₂O₆S₂K₂
MW: 378.55 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
CAS Number: 108321-27-3
EINECS: 227-057-6
Merck Index: 12,07633

Order Now

Specification Test Results

Heavy Metals = 15 ppm
Loss on Drying = 2%
pH (1%, Water) @25°C 9.5 - 10.5
Purity = 97%
Purity = 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR