Barium chloride dihydrate ≥99% ACS

Danger

VWRVE418-500G
89230-010CS 263.86 USD
89230-010
Barium chloride dihydrate ≥99% ACS
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
MW: 244.27 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3.86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 13,00974

Order Now

Specification Test Results

Assay 99%
Calcium = 0.05%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.005%
Iron = 0.0002%
Loss on Drying 14 - 16%
Oxidizing Substances (as NO3) = 0.005%
pH (5%, Water) @25°C 5.2 - 8.2
Potassium = 0.0025%
Sodium = 0.005%
Strontium = 0.1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR