Sodium thiocyanate, Reagent Grade

Warning

Synonyms: Sodium rhodanide

VWRV0861-500G
10118-144EA 59.75 USD
10118-144
Sodium thiocyanate, Reagent Grade
Sodium thiocyanate
Formula: NaSCN
MW: 81.07 g/mol
Melting Pt: 287 °C
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011123
CAS Number: 540-72-7
EINECS: 208-754-4
Merck Index: 12,09463

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR