Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tube Connector Fittings, S.C.A.T

Supplier: VWR INTERNATIONAL GMBH
Exchanging fittings will increase the number of connections or connect tubes with different diameters.


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Material For Tube Diameter Supplier No. VWR Catalog Number Unit Price Quantity
Triple Collector, Straight PTFE/PP 3 x 6.4 mm 160128 10060-378 Each Retrieving
Tube Fitting, Curved PTFE 6.4 - 8.0 mm 160143 10060-342 Each Retrieving
Tube Fitting, Curved PP 6.4 - 9 mm 117808 10060-340 Each Retrieving
Tube Fitting, Straight PP 2.0 - 3.0 mm 107811 10060-346 Each Retrieving
Tube Fitting, Straight PP 4.0 - 6.0 mm 107813 10060-350 Each Retrieving
Tube Fitting, Straight PP 6.2 - 7.5 mm 117816 10060-338 Each Retrieving
Tube Fitting, Straight PTFE 6.4 - 8.0 mm 160142 10060-344 Each Retrieving
Tube Fitting, Straight PP 9.5 - 11.0 mm 107817 10060-354 Each Retrieving
Tube Fitting, Straight PP 3.0 - 4.0 mm 107812 10060-348 Each Retrieving
Tube Fitting, Straight PP 5.0 - 7.0 mm 107814 10060-352 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty