Would you like to log in again?
Puritan® Polyester Tipped Swab, Puritan Medical Products
Supplier: PURITAN MEDICAL PRODUCTS COMP
25-800D50
95041-315CS 517.15 USD InStock
95041-315
Puritan® Polyester Tipped Swab, Puritan Medical Products
Swabs Swabs for Sampling
These synthetic applicator tips are made of medical-grade polyester fiber carded in our factory
The thin flexible aluminum handle follows contours of passages with virtually no injury to mucosal tissues. Applications to include forensic evidence collection, diagnostic specimen collection for testing and screening, flu and strep testing, pediatric, and eye microbiology.
They contain no fatty acids or latent sugars, making them ideal for specimen transport, lab work, and diagnostic kits.
Certifications: CE marked.
Ordering information: Made in the USA.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty