General Purpose Circulating Refrigeration Units with External Pump, Grant Instruments
Supplier: Grant
TC120-R4 US
10067-684EA 11620.34 USD
10067-684
General Purpose Circulating Refrigeration Units with External Pump, Grant Instruments
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The Grant Optima™ TC120 R series of refrigerated baths and circulators with digital control provides excellent value for money for more demanding applications requiring accurate heating and cooling in the range of -25 to 100°C
The TC120 immersion thermostat incorporates Intelligent Control Optimization (ICO)™ for adaptive intelligent PID temperature control and includes a powerful integral pump (maximum flow rate 16L/min), making TC120 series refrigerated systems suitable both for immersing samples and circulating temperature controlled fluid to external devices. It includes a convenient timer function for reaction timing and a variable high temperature alarm setting. The temperature range and heater power are automatically limited according to the liquid type selected.
All three units contain a simple-to-use rotor dial with two function keys providing access to the interactive interface for fast, accurate set-up. They have a clear 4-digit display for instant reassurance which is easy to read from a distance and a visual alarm which alerts you when your attention is required.
Combine the versatile Grant Optima™ digital immersion thermostat TC120 with a choice of three Grant refrigeration (R) units. Choose from a 5L or 20L tank and from a range of cooling capacities.
Adjustable overtemperature cut out. External circulation.
Storage capacity: Three temperature values
Certifications: CE marked.
Ordering information: The refrigeration unit can be switched off independently of the thermostat to allow heating only applications, except for the R1 unit without relay control. All the refrigeration base units can be used up to a maximum temperature of 100°C.