Advanced Circulating Refrigeration Units with External Pump, Grant Instruments
Supplier: Grant
TX150-R4 US
10067-652EA 8931.82 USD
10067-652
Advanced Circulating Refrigeration Units with External Pump, Grant Instruments
The Grant Optima™ TX150 series of digitally controlled refrigerated circulating baths provides outstanding performance for routine and sophisticated applications requiring accurate temperature control in the range of -47 to 100°C
The Grant Optima™ TX150 is a high-performance programmable digital heating circulator with a powerful pump for external circulation (maximum flow 18 L/min) an exceptionally simple user interface. This allows fast set-up of multiple parameters through a powerful menu system and provides sophisticated control for temperature-controlled applications.
The TX150 has USB connectivity to a PC or laptop with memory capacity for one program of up to 30 segments, and a choice of five languages; English, French, German, Spanish and Italian. It is programmable with Labwise™, a software program to set all parameters, including programs, via the PC. Labwise™ also features real-time status windows, a graphic display of the controller's performance as a program is run, and allows data to be logged for future recall and analysis. Full color QVGA TFT display.
Combine the versatile Grant Optima™ digital immersion thermostat TX150 with a choice of four Grant refrigeration units. Choose from a 5L or 20L tank and from a range of cooling capacities.
The series refrigerated systems combine the powerful and versatile Grant Optima™ digital heating circulator TX150 with a choice of four stainless steel refrigeration units. Offset adjustment. Socket for external probe. Programmable remote via PC. Relay control. Adjustable overtemperature cut out. Drain tap. External circulation. One relay.
Certifications: CE Marked.
Ordering information: The refrigeration unit can be switched off independently of the thermostat to allow heating only applications, except for the R1 unit without relay control. All the refrigeration base units can be used up to a maximum temperature of 100°C.