Mouse Recombinant GACRP30 (from E. coli)
Supplier: ProSci
40-487
10072-588EA 302.43 USD
10072-588
Mouse Recombinant GACRP30 (from E. coli)
Proteins and Peptides
gAcrp30 is a naturally occurring globular protein, obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, gAcrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and names AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver. Recombinant murine gAcrp30 is a 16.6 kDa protein consisting of 145 amino acid residues.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.