Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

RELM alpha Recombinant Protein, ProSci Inc

Supplier: Prosci
40-484
RELM-alpha belongs to a unique family of tissue-specific cytokines termed FIZZ (found in inflammatory zone) and RELM. The three known members of this family; Resistin, RELM-alpha and RELM-beta are 85-94 amino acid secreted proteins sharing a conserved C-terminal domain characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C. RELM-alpha and Resistin are secreted exclusively by adipocytes while RELM-beta is expressed in the epithelium of the colon and small bowel. The physiological role and molecular targets of RELM-alpha re still unknown. Recombinant murine RELM-alpha is a 10.0 kDa monomeric protein containing 88 amino acid residues.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.005 mg 40-484 10072-584 Each (5µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name RELM alpha
Protein Synonyms RELMalpha, Cysteine-rich secreted protein A12-gamma, Xcp2, Fizz1, 1810019L16Rik, HIMF, Himf, Pmng1, RELM-alpha, Resistin-like alpha, Fizz-1, RELMa
Protein/Peptide Type Recombinant
Species Mouse
Source E. coli
Gene ID 57262
Purity >98%
Formulation Lyophilized
Protein Type Recombinant
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty