Would you like to log in again?
Beveled Orifice Pipet Tips, 1–200 µL
Supplier: Thermo Scientific
110-96RB
An excellent fitting pipet tip is one that allows the pipettor to reach its peak performance." Our goal is to provide the best fitting tips in the marketplace
Today, there are a myriad of pipettors to consider when designing and manufacturing pipet tips. Because dimensions and applications vary from one pipettor to the next, we offer a variety of pipet tips to select from. With more than fifty pipet tips, covering a volume range of 0.1 to 10,000µL, available with or without filters, in a variety of colors, materials, and packaging configurations, the researcher can select the best-fitting tips for their particular pipettor and application. These pipet tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product. A mold release agent is NOT used in the manufacturing of these pipet tips. Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals. A trace metal certificate is available upon request. Certified Pipet Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free (non-sterilized product is not certified as pyrogen-free.) Certificate of analysis is available upon request. Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation. Certificate of irradiation available upon request.
ORDER
Expand All / Collapse All
Description Volume Tip Color Packed Supplier No. VWR Catalog Number Unit Price Quantity
Beveled Orifice Tips 1–200 µL Yellow 96 Tip Rack with Lift-Off Lid 110-96RB 490016-502 Box of 960 Retrieving
Case of 4800 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty