NanoRam Handheld RAMAN System, B&W Tek
840000306 840000364 840000344 840000397 840000351 840000365 840000354 840000355 840000348 840000398 840000343 840000366 840000307 840000319 840000305 840000295 840000342 840000128 840000291 840000290 840000318 840000379 840000301 840000316 840000350 840000345 840000352 840000341 840000298 840000372 840000361 840000371 840000353 840000347 840000309 840000349 840000294 840000296 840000308 840000329 840000370 840000320 840000399
10121-536EA 91.43 USD
10121-536 10121-514 10121-516 10121-538 10121-478 10121-532 10121-510 10121-512 10121-534 10121-474 10121-496 10121-550 10121-476 10121-530 10121-552 10121-492 10121-470 10121-494 10121-472 10121-490 10121-508 10121-504 10121-548 10121-526 10121-506 10121-528 10121-544 10121-500 10121-522 10121-488 10121-502 10121-546 10121-524 10121-468 10121-540 10121-484 10121-462 10121-542 10121-520 10121-486 10121-480 10121-482 10121-518
NanoRam Handheld RAMAN System, B&W Tek
The NanoRam is a state-of-the-art handheld RAMAN instrument for nondestructive identification and verification of materials such as APIs, excipients, intermediates and finished products
RAMAN spectroscopy is an approved method by the U.S. and European Pharmacopoeia, as well as the Pharmacopoeia of the People’s Republic of China. It is also a well-recognized method for compliance with the PIC/S GMP guide regarding 100% identity assurance for starting materials. The NanoRam is fully compliant with all governing regulations, including 21 CFR Part 11 and Part 1040.10, and can play an integral role in cGMP compliant facilities. B&W Tek offers a wide variety of services, including assistance with method and/or new library development as well as support with IQ/OQ/PQ implementation.
Features include high-brightness touch screen display, embedded 1 and 2 dimensional barcode scanner, IP-64 dust tight and splash proof rated housing, sampling accessories for almost any environment, batch scanning option for large volume operation, intuitive software for technical and non-technical users, Wi-fi and Ethernet communication for data sync and management, and library and method transfer capabilities.
Compact and agile, the NanoRam can be used by non-technical users to rapidly identify samples in the lab, warehouse, loading dock or field, helping to eliminate quarantine areas and expedite materials through the manufacturing lifecycle. Utilizing RAMAN technology, non-contact analysis can be performed through transparent containers, all while maintaining the volume and integrity of the sample.
Contact your VWR Sales Representative for more information, to request a quote or set up a demo.
Certifications: 21CFR part 11; CDRH 1040.10; IP-64; USP Chapter 1120.
Ordering information: The NanoRam Handheld RAMAN Spectrometer comes standard with a 24 month warranty and the following items: a power adaptor, one rechargeable battery, a point and shoot adaptor, a liquid vial holder adaptor, a bottle adaptor, a pouch for adaptors, a pack of 15mm-diameter glass vials, a pouch for the 15mm vials, a calibration cap with built-in ASTM standard polystyrene, one pair of laser safety goggles, one shaft protection cap, a stylus/pen combo for NanoRam touch screen, EU, UK and US country specific power plug adaptors, a LAN cable, and hard copies of User Manual and NanoRam ID Installation Guide. All items are packaged and shipped in a rugged carrying case.