Triple Mix Paddle Blender, Boekel Scientific
Supplier: Boekel Scientific
321500 321500-2
10040-414EA 8323.54 USD
10040-414 10040-416
Triple Mix Paddle Blender, Boekel Scientific
Homogenizers Laboratory Paddle Blenders
Blender is designed for use in food safety testing and macro sample preparation applications
The large process viewing window and easy-to-use digital control system allows users to view the sample status without opening the door. The unit features an easy to clean design with removable paddles and drip tray. To facilitate use in high throughput labs, an Auto-Run feature is included, allowing users to set and repeat the same times and rpm's in a continuous fashion.
The unit is designed to be used with standard and filter-type food sample preparation bags. The unique three paddle design provides excellent homogenization and mixing.
Certifications: The 115V unit is UL and CSA certified. Both 115V and 230V units are CE listed.
Ordering information: Unit includes a two-year manufacturer's warranty.