Human Recombinant HCC1 (from E. coli)
Supplier: ProSci
40-391
10072-172EA 302.43 USD
10072-172
Human Recombinant HCC1 (from E. coli)
Proteins and Peptides
HCC-1 is a CC chemokine that signals through the CCR1 receptor and chemoattracts blood monocytes. It is secreted by various tissues including skeletal muscle, heart, spleen, liver, bone marrow and plasma. Mature HCC-1 is found in four different forms, which are distinguished by differential N-terminal truncation and contain 74, 72, 71, or 66 amino acid residues. Recombinant human HCC-1 (66 a.a.) is a 7.8 kDa protein consisting of 66 amino acids including the four highly conserved residues present in CC chemokines.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.