Anti-SNCB Chicken Polyclonal Antibody

Supplier: ProSci
XW-8053
10071-404EA 948.1 USD
10071-404
Anti-SNCB Chicken Polyclonal Antibody
Antibodies
The protein encoded by this gene is highly homologous to beta-synuclein. These proteins are abundantly expressed in the brain and putatively inhibit phospholipase D2 selectively. The encoded protein, which may play a role in neuronal plasticity, is abundant in neurofibrillary lesions of patients with Alzheimer disease. This protein has been shown to be highly expressed in the substantia nigra of the brain, a region of neuronal degeneration in patients with Parkinson disease; however, no direct relation to Parkinson disease has been established. Two transcript variants encoding the same protein have been found for this gene.

SNCB protein antibody can be used for the detection of SNCB protein by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SNCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR