Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VWF Rabbit Polyclonal Antibody

Supplier: Prosci

70-907
10080-382EA 530.67 USD InStock
10080-382
Anti-VWF Rabbit Polyclonal Antibody
Antibodies
VWF / von Willebrand Factor Antibody can be used for Immunofluorescencestarting at , Immunohistochemistry in frozen tissues and Immunohistochemistry in paraffin embeded tissues starting at 5μg/mL.

Purified

PBS, pH 7.4, 15mM sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: VWF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty