Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

Anti-GHRL Rabbit Polyclonal Antibody

Supplier: Prosci

70-906
10080-380EA 530.67 USD InStock
10080-380
Anti-GHRL Rabbit Polyclonal Antibody
Antibodies
GHRL / Ghrelin Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 - 20 μg/mL.

Protein A Column

PBS, pH 7.2, 30% glycerol, 0.5% BSA, 5 mM EDTA and 0.01% thimerosal.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GHRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty