Anti-CD79a Rabbit Polyclonal Antibody

Supplier: ProSci
70-899
10080-366EA 705.02 USD
10080-366
Anti-CD79a Rabbit Polyclonal Antibody
Antibodies
Human IgA Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5.

10mM PBS, pH 7.4, with 0.2% BSA and 0.09% Sodium Azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD79A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR