Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FANCC Rabbit Polyclonal Antibody

Supplier: Prosci

70-887
10074-094EA 530.67 USD InStock
10074-094
Anti-FANCC Rabbit Polyclonal Antibody
Antibodies
FANCC Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 μg/mL, and Western Blotting.

Affinity Purified

PBS, pH 7.2, without preservatives.

Antibody should be stored at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: FANCC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty