Anti-LEO1 Rabbit Polyclonal Antibody

Supplier: ProSci
70-874
10074-068EA 694.79 USD
10074-068
Anti-LEO1 Rabbit Polyclonal Antibody
Antibodies
LEO1 Antibody can be used for ELISA starting at 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LEO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR