Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RPS27A Rabbit Polyclonal Antibody

Supplier: Prosci

70-873
10074-066EA 530.67 USD InStock
10074-066
Anti-RPS27A Rabbit Polyclonal Antibody
Antibodies
Ubiquitin +1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 μg/mL, and Western Blotting starting at 1:5000.

Protein A Column

HEPES, 0.15 M NaCl, 0.01% BSA, 0.03% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RPS27A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty