Anti-GC Rabbit Polyclonal Antibody

Supplier: ProSci
70-872
10074-064EA 719.12 USD
10074-064
Anti-GC Rabbit Polyclonal Antibody
Antibodies
GC / VDBP Antibody can be used for ELISAstarting at , and Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR