Anti-STEAP2 Rabbit Polyclonal Antibody

Supplier: ProSci
70-810
10080-278EA 772.82 USD
10080-278
Anti-STEAP2 Rabbit Polyclonal Antibody
Antibodies
STEAP2 Antibody can be used for ELISAstarting at , and Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STEAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Dog, Horse, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR