Anti-MMP2 Rabbit Polyclonal Antibody

Supplier: ProSci
70-803
10080-266EA 505.97 USD
10080-266
Anti-MMP2 Rabbit Polyclonal Antibody
MMP2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 20 ?g/mL, and Western Blotting starting at 5 - 10 ?g/mL.

10mM PBS, pH 7.4, with 0.2% BSA and 0.09% Sodium Azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR