Anti-CLDN7 Rabbit Polyclonal Antibody

Supplier: ProSci
70-797
10080-254EA 694.79 USD
10080-254
Anti-CLDN7 Rabbit Polyclonal Antibody
Antibodies
CLDN7 / Claudin-7 Antibody can be used for Immunohistochemistry starting at 1:25.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CLDN7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR