Anti-DYSF Rabbit Polyclonal Antibody

Supplier: ProSci
70-776
10080-212EA 719.12 USD
10080-212
Anti-DYSF Rabbit Polyclonal Antibody
Antibodies
DYSF / Dysferlin Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:50, and Western Blotting starting at 1 ?g/mL.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DYSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR