Anti-CDK1 Rabbit Polyclonal Antibody

Supplier: ProSci
70-748
10075-160EA 505.97 USD
10075-160
Anti-CDK1 Rabbit Polyclonal Antibody
CDK1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:50, Immunoprecipitation and Western Blotting starting at 1:25.

10 mM PBS, pH 7.4, BSA, sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDK1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR