Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NSF Rabbit Polyclonal Antibody

Supplier: Prosci

70-735
10075-134EA 530.67 USD InStock
10075-134
Anti-NSF Rabbit Polyclonal Antibody
Antibodies
NSF Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 μg/mL, Immunoprecipitation and Western Blotting.

Protein A Column

PBS, 50% glycerol, 1% BSA, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty